Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EMAP-II/AIMP1 Mouse anti-Human, Rat, Clone: 546-2, Novus Biologicals™
SDP

Catalog No. NB119902 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB119902 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB119902 Supplier Novus Biologicals Supplier No. NBP311277

Mouse Monoclonal Antibody

EMAP-II/AIMP1 Monoclonal specifically detects EMAP-II/AIMP1 in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry-Paraffin, Immunoassay.

Specifications

Antigen EMAP-II/AIMP1
Applications Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin), Immunoassay
Classification Monoclonal
Clone 546-2
Concentration 0.1 mg/ml
Conjugate Unconjugated
Dilution Western Blot, Flow Cytometry, Immunohistochemistry-Paraffin, Immunoassay
Formulation 0.2 um filtered solution in PBS, 0.1% BSA
Gene Alias aminoacyl tRNA synthetase complex-interacting multifunctional protein 1, EMAP-2, EMAPIImember 1 (endothelialmonocyte-activating), endothelial-monocyte activating polypeptide II, Multisynthase complex auxiliary component p43, multisynthetase complex auxiliary component p43
Host Species Mouse
Immunogen Monoclonal antibody 546-2 reacts with endothelial-monocyte-activating polypeptide II (EMAP II).
Purification Method Protein G purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 9255
Target Species Human, Rat
Content And Storage Store at 4°C. Do not freeze.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.