Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EMAP II Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598413
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598413 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598413 Supplier Invitrogen™ Supplier No. PA598413
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

EMAP II Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

The protein encoded by this gene is a cytokine that is specifically induced by apoptosis, and it is involved in the control of angiogenesis, inflammation, and wound healing. The release of this cytokine renders the tumor-associated vasculature sensitive to tumor necrosis factor. The precursor protein is identical to the p43 subunit, which is associated with the multi-tRNA synthetase complex, and it modulates aminoacylation activity of tRNA synthetase in normal cells. This protein is also involved in the stimulation of inflammatory responses after proteolytic cleavage in tumor cells. Multiple transcript variants encoding different isoforms have been found for this gene. A pseudogene has been identified on chromosome 20.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EMAP II
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 3.68 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene Aimp1
Gene Accession No. Q12904
Gene Alias 9830137A06Rik; aaRS-interacting multifunctional protein 1; AIMP1; AIMP1/p43; AIMP1-PA; Aminoacyl tRNA synthase complex-interacting multifunctional protein 1; aminoacyl tRNA synthetase complex interacting multifunctional protein 1; aminoacyl tRNA synthetase complex-interacting multifunctional protein 1; aminoacyl-tRNA synthetase-interacting multifunctional protein 1; ARS inducible multifunctional protein 1; ARS-interacting multifunctional protein 1; CG8235; CG8235-PA; Dmel\CG8235; Dmel_CG8235; EMAP2; EMAP-2; EMAPII; EMAP-II; endothelial monocyte activating polypeptide 2; endothelial monocyte-activating polypeptide 2; Endothelial monocyte-activating polypeptide II; endothelial-monocyte activating polypeptide II; HLD3; mca1; multisynthase complex auxiliary component p43; multisynthetase complex auxiliary component p43; p43; SCYE1; Small inducible cytokine subfamily E member 1; small inducible cytokine subfamily E, member 1; small inducible cytokine subfamily E, member 1 (endothelial monocyte-activating)
Gene Symbols Aimp1
Host Species Rabbit
Immunogen Recombinant Human Aminoacyl tRNA synthase complex-interacting multifunctional protein 1 protein (25-180AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9255
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.