Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Embigin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA523257
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA523257 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA523257 Supplier Invitrogen™ Supplier No. PA523257
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Embigin Polyclonal Antibody for Western Blot

Embigin is a 327 amino acid regulatory protein belonging to the immunoglobulin superfamily class of CAMs, and has two Ig-like (immunoglobulin-like) V- type domains. In situ hybridization analysis shows that Embigin has specific carbohydrate epitopes and plays an important role in early postimplantation embryogenesis. A highly glycosylated transmembrane glycoprotein, Embigin acts as a regulator of cell/ECM interactions during development and homeostasis of normal adult tissues. Transfection studies suggest its role in promotion of integrin-mediated cell-substratum adhesion. Widely expressed in most of the tissues and embryonal carcinoma cells, prostate and mammary cancer cell lines, it may also help in the classification of human carcinomas.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Embigin
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA and 0.05% sodium azide
Gene EMB
Gene Accession No. P21995, Q6PCB8
Gene Alias AL022799; EMB; Embigin; embigin homolog; glycoprotein 70; Gp70; Teratocarcinoma glycoprotein Gp-70
Gene Symbols EMB
Host Species Rabbit
Immunogen A portion of amino acids 130-180 of human Embigin.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 133418, 13723
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.