Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EMC7 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP318408
This item is not returnable.
View return policy
Description
EMC7 Polyclonal antibody specifically detects EMC7 in Human, Mouse samples. It is validated for Western BlotSpecifications
EMC7 | |
Polyclonal | |
Unconjugated | |
Tris-citrate/phosphate pH 7 -8. | |
Rabbit | |
Affinity purified | |
RUO | |
56851 | |
Store at 4°C. Do not freeze. | |
IgG |
Western Blot | |
1 mg/ml | |
Western Blot 1:2000 - 1:10000 | |
C11orf3, C15orf24, chromosome 15 hypothetical ATG/GTP binding protein, chromosome 15 open reading frame 24, ER membrane protein complex subunit 7, HT022, ORF1-FL1 | |
Recognizes region between residues Between Between 1 and 50.. | |
100 μg | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction