Learn More
Invitrogen™ EMC7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113896
Description
Immunogen sequence: VDITSKGKMR ARYVNYIKTS EVVRLPYPLQ MKSSGPPSYF IKRESWGWTD For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
EMC7 (ER Membrane Protein Complex Subunit 7) is a Protein Coding gene. Diseases associated with EMC7 include Cone-Rod Dystrophy, X-Linked, 1. Gene Ontology (GO) annotations related to this gene include carbohydrate binding. [GeneCards].
Specifications
| EMC7 | |
| Polyclonal | |
| Unconjugated | |
| EMC7 | |
| 2900064A13Rik; AI451465; c11orf3; C15orf24; chromosome 15 hypothetical ATG/GTP binding protein; Emc7; ER membrane protein complex subunit 7; HT022; ORF1-FL1; Orf3; UNQ905/PRO1926; UPF0480 protein C15orf24; UPF0480 protein C15orf24 homolog | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 56851 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9NPA0 | |
| EMC7 | |
| Synthetic peptide directed towards the middle region of human C15orf24. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.