Learn More
MilliporeSigma™ Accessories for XX2702550 1225 Sampling Manifold
Catalog No. p-109521 Shop All MilliporeSigma Products
Click to view available options
Type:
Ball Valve
Accessories for filter holders for sample preparation.
Accessories for filter holders for sample preparation.
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
Connectors 1/4 in. Stepped hose connector / 1/4 in. NPTM
For Use With (Equipment) 1225 sampling manifold
Material Glass-filled Polypropylene
Type Ball Valve
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.