Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ α2-3,6,8,9-Neuraminidase, Arthrobacter ureafaciens, Recombinant Protein, E. coli
Shop All MilliporeSigma Products
Click to view available options
Quantity:
250 mIU
Description
Recombinant, Arthrobacter ureafaciens α2-3,6,8,9-Neuraminidase expressed in E. coli. Catalyzes the hydrolysis of all non-reducing terminal sialic acid residues from complex carbohydrates and glycoproteins. High concentrations of this sialidase and prolonged periods of incubation result in the additional cleavage of branched sialic acids. This enzyme is found as two active species of MW 60000 and 69000.

Specifications
Specifications
Formulation | In 20mM Tris-HCl, 20mM NaCl, pH 7.5. |
Quantity | 250 mIU |
Source | E. coli |
Immunogen | α2-3,6,8,9-Neuraminidase |
Regulatory Status | RUO |
Species | A. ureafaciens |
Recombinant | Recombinant |
Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction