Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ β2-Microglobulin, Human Urine, Protein
Shop All MilliporeSigma Products
Click to view available options
Quantity:
100 μg
250 μg
Description
Native β2-microglobulin from human urine. A useful marker for the determination of AIDS progression in HIV-infected subjects. Increased levels may also indicate increased proliferation of tumor masses.
Specifications
Specifications
| Formulation | In PBS, 150mM NaCl, 20mM phosphate buffer, pH 7.3. |
| Purification Method | SDS-PAGE |
| Quantity | 250 μg |
| Immunogen | β2-Microglobulin |
| Storage Requirements | Following initial thaw, product can be stored at 4°C for up to 6 months. |
| Regulatory Status | RUO |
| Species | Human |
| Recombinant | Native |
| Form | Liquid |
| Purity or Quality Grade | ≥98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction