Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ β2-Microglobulin, Human Urine, Protein

Catalog No. p-5365158 Shop All MilliporeSigma Products
Click to view available options
Quantity:
100 μg
250 μg
Native β2-microglobulin from human urine. A useful marker for the determination of AIDS progression in HIV-infected subjects. Increased levels may also indicate increased proliferation of tumor masses.
TRUSTED_SUSTAINABILITY

Specifications

Formulation In PBS, 150mM NaCl, 20mM phosphate buffer, pH 7.3.
Purification Method SDS-PAGE
Quantity 250 μg
Immunogen β2-Microglobulin
Storage Requirements Following initial thaw, product can be stored at 4°C for up to 6 months.
Regulatory Status RUO
Species Human
Recombinant Native
Form Liquid
Purity or Quality Grade ≥98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.