Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Amicon™ Pro Purification System with Ultra-0.5 Device, 3 Kda
Shop All MilliporeSigma Products
Click to view available options
Quantity:
12Pack
24Pack
Includes:
12 Amicon Ultra NMWCO 3 kDa filters, 12 Exchange devices, and 12 Collection tubes with cap
24 Amicon Ultra NMWCO 3 kDa filters, 24 Exchange devices, and 24 Collection tubes with cap
Description
The Amicon™ Pro Purification System includes: Amicon Ultra NMWCO filters, Exchange devices, and Collection tubes with cap.
Specifications
Specifications
| Disposable | Single-use |
| MWCO | 3 kDa |
| Sterility | Non-sterile |
| Housing Material | Regenerated Cellulose (RC) |
| Includes | 12 Amicon Ultra NMWCO 3 kDa filters, 12 Exchange devices, and 12 Collection tubes with cap |
| Quantity | 12Pack |
| Type | Centrifugal Filters |
| Volume (Metric) | 15 mL |
| Length (Metric) | 12 cm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction