Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Atrial Natriuretic Factor 1-28, Human Neuropeptide, Calbiochem
Shop All MilliporeSigma Products
Click to view available options
Quantity:
1 mg
Description
Increases the rate of renal excretion. Higher levels are reported in congestive heart failure. Inhibits MAP kinase through the natriuretic peptide clearance receptor.
Specifications
Specifications
| Species | Human |
| Purification Method | HPLC |
| Purity | ≥97% |
| CAS | 91917-63-4 |
| Color | White to Off-white |
| Content And Storage | Following reconstitution, aliquot and freeze (−20°C). Stock solutions are stable for up to 1 month at −20°C. |
| Format | Lyophilized Solid |
| Formulation | Supplied as a trifluoroacetate salt. |
| Product Type | Atrial Natriuretic Factor 1-28 |
| Quantity | 1 mg |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction