Learn More
Specifications
Specifications
| Content And Storage | Room temperature in clean dry area away from any chemicals or direct light |
| Description | AXIS Axon Isolation Device, Tissue Culture Ready |
| For Use With (Application) | For Cell Culture |
| Includes | 10 units of AX450, axon isolation device consisting of four wells, two channels, and set of microgrooves |
| Quantity | 10 Devices |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.