Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ BrdU Cell Proliferation Assay Kits
Click to view available options
Quantity:
1000 Tests
200 Tests
Description
BrdU Cell Proliferation Assay is a non-isotopic immunoassay for quantification of BrdU incorporation into newly synthesized DNA of actively proliferating cells. It is sensitive, rapid, and easy to perform.
Kit Components:
- BrdU Label 58
- Fixative/Denaturing Solution 58
- Anti-BrdU Antibody 58
- Antibody Diluent 58
- TMB (Soluble)
- ELISA Conjugate Diluent(-20C, 4%BSA)
- ELISA Stop Solution
- ELISA 20X Plate Wash Concentrate
- Peroxidase Goat anti-Mouse IgG

Specifications
Specifications
Format | 96-well plate |
Content And Storage | Store at -20°C. Avoid freeze/thaw cycles. |
Quantity | 200 Tests |
For Use With (Application) | Non-isotopic immunoassay for quantification of BrdU incorporation into newly synthesized DNA of actively proliferating cells |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction