Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ FASTsilver™ Gel Staining Kit

Catalog No. 3412981kit
Click to view available options
Quantity:
1 Kit

Rapid and sensitive method for detecting protein and nucleic acids fractionated by PAGE

  • Provides gels with clear backgrounds and sharp protein or nucleic acid image
  • Silver staining is about 100 times more sensitive than the standard Coomassie Blue protein staining and about 10 times more sensitive than ethidium bromide for DNA and RNA
TRUSTED_SUSTAINABILITY

Specifications

Color Silver
Product Type DNA/RNA/Protein Staining Kit
Quantity 1 Kit
Storage Requirements 15° to 30°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.