Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ PANSORBIN™ Cells, Lyophilized

Catalog No. p-5267540 Shop All MilliporeSigma Products
Click to view available options
Quantity:
1 g
5 g

Catalog No. 50-786-21GM

Supplier: MilliporeSigma™ 5078621GM

Only null left
Add to Cart
Add to Cart

Lyophilized cells for enzyme immunoassay, immunoprecipitation and radioimmunoassay

PANSORBIN cells are formalin-fixed and heat-killed Staphylococcus aureus cells bearing protein A that bind to the Fc portion of human IgG subclasses 1, 2, and 4, and to IgG from other species.

  • Form: Lyophilized
  • Formulation: Lyophilized from PBS, 0.1% NaN3
  • Reconstitution: Reconstitute in sterile distilled H2O
  • Soluble in sterile dH2O
  • Applications: Enzyme Immunoassay, Immunoprecipitation, Radioimmunoassay
  • Storage: Store at 2° to °C

TRUSTED_SUSTAINABILITY

Specifications

Product Type PANSORBIN Cells
Content And Storage 2°C to 8°C
Quantity 1 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.