Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ PANSORBIN™ Cells, Standardized

Catalog No. p-5267541 Shop All MilliporeSigma Products
Click to view available options
Quantity:
25 mL
50 mL

For mitogenic stimulation of B lyphocytes and for immunoprecipitation

PANSORBIN cells are useful as a solid-phase IgG-binding reagent due to the high-affinity interaction of protein A with the Fc domain of IgG. They work better when the antibody is human (IgG1, IgG2, IgG4), rabbit IgG (all isotypes), or mouse (IgG2a, IgG2b, IgG3).

  • Activity: Binding capacity: ≥2mg/mL
  • Form: Liquid
  • Formulation: Supplied as a 7.5 to 15% (w/v) Staphylococcus aureus cell suspension in PBS, 0.1% sodium azide, pH 7.2
  • Preservative: 0.1% sodium azide
  • Storage: Store at 2° to 8°C

TRUSTED_SUSTAINABILITY

Specifications

Product Type PANSORBIN Cells
Content And Storage 2°C to 8°C
Quantity 50 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.