Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calmodulin, Bovine Brain, High Purity, Protein
Shop All MilliporeSigma Products
Click to view available options
Quantity:
1 mg
5 mg
Description
A ubiquitous Ca2+-binding protein that serves as a physiological effector of a wide range of biological processes. Calmodulin highly purified from bovine brain.
Specifications
Specifications
| Formulation | Lyophilized from 30mM HEPES, 1mM CaCl2, 0.1mM DTT, pH 7.4. |
| Preparation Method | Aqueous buffers or H2O. Please refer to the vial label for lot-specific reconstitution volume. |
| Purification Method | SDS-PAGE |
| Quantity | 1 mg |
| Immunogen | Calmodulin |
| Storage Requirements | Following reconstitution, aliquot and freeze (−20°C or −70°C). Aqueous stock solutions are stable for up to 2 months at −20°C or for up to 4 months at −70°C. |
| Regulatory Status | RUO |
| Species | Bovine |
| Recombinant | Native |
| Form | Lyophilized |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction