Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Casein, Bovine Milk, Carbohydrate and Fatty Acid Free, Protein

Catalog No. p-5364942 Shop All MilliporeSigma Products
Click to view available options
Quantity:
100 g
500 g

Catalog No. 21-868-2500GM

Supplier: MilliporeSigma™ 218682500GM

Only null left
Add to Cart
Add to Cart
High molecular weight phosphoprotein. Useful as a substrate for bromelain, chymotrypsin, pepsin, trypsin, and other proteases. Supplied as Hammarsten grade material.
TRUSTED_SUSTAINABILITY

Specifications

Preparation Method Dilute Alkaline Solutions (10mg/mL)
Purification Method Assay
Quantity 500 g
Immunogen Casein
Storage Requirements Following reconstitution, store in the refrigerator (4°C). Stock solutions are stable for up to 1 week at 4°C. After one week, it is recommended that fresh solution be prepared.
Regulatory Status RUO
Without Additives Carbohydrate and Fatty Acid Free
Species Bovine
Recombinant Native
Form Solid
Purity or Quality Grade ≥95%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.