Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Casein, Bovine Milk, Protein

Catalog No. 218680100GM Shop All MilliporeSigma Products
Click to view available options
Quantity:
25 g
100 g
High molecular weight phosphoprotein. Useful as a substrate for bromelain, chymotrypsin, pepsin, trypsin, and other proteases.
TRUSTED_SUSTAINABILITY

Specifications

Preparation Method Dilute alkaline solutions (1mg/mL)
Quantity 100 g
Immunogen Casein
Storage Requirements Following reconstitution, aliquot and freeze (−20°C). Stock solutions are stable for up to 3 months at −20°C.
Regulatory Status RUO
Species Bovine
Recombinant Native
Form Solid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.