Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Cathepsin S, Human, Recombinant Protein, E. coli

Catalog No. 21934325UG Shop All MilliporeSigma Products
Click to view available options
:
25μg

Catalog No. 21-934-325UG

Supplier: MilliporeSigma™ 21934325UG

Only null left
Add to Cart
Add to Cart
Recombinant, human cathepsin S expressed in E. coli without a tag or fusion protein. A major lysosomal cysteine proteinase preferentially expressed in macrophages and microglia. Exhibits high specific activity.
TRUSTED_SUSTAINABILITY

Specifications

Formulation In 35mM potassium phosphate, 35mM Sodium acetate, 2mM DTT, 2mM EDTA, 50% ethylene glycol, pH 6.5.
Purification Method SDS-PAGE
Quantity 25μg
Source E. coli
Immunogen Cathepsin S
Storage Requirements Following initial thaw, aliquot and freeze (−70°C).
Regulatory Status RUO
Species Human
Recombinant Recombinant
Form Liquid
Purity or Quality Grade ≥90%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.