Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Certistain™ Rhodamine B Dye
Click to view available options
Quantity:
100g
Description
Rhodamine B (C.I. 45170) is a dry dye for fluorescence Tb staining of bacteriological smears and histological tissue sections of human origin.The mechanism is a non-specific binding of fluorescence dye on a substrate, similar to the classical Ziehl-Neelsen method, although the heating of the Rhodamine B color solution can be avoided. The dye is a CE certified IVD product and has a constant dye content and a reliable high quality, which is one of the major precondition for a reproducible diagnostic staining.

Specifications
Specifications
Quantity | 100g |
Type | Rhodamine B Dye |
For Use With (Application) | Fluorescence Tb staining of bacteriological smears, histological tissue sections of human origin |
Melting Point | 199-201°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction