Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Chemicon™ AOF ITS Supplement (100x)

Catalog No. p-5235437 Shop All MilliporeSigma Products
Click to view available options
Quantity:
10 mL
100 mL

an animal-origin-free product that contains human recombinant insulin, transferrin along with sodium selenite and is used for the serum-free culture of numerous cell types.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2-8°C
For Use With (Application) Stem cell culture, cell culture, cell stimulation, cell growth
Quantity 100 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.