Learn More
MilliporeSigma™ Chemicon™ ApopTag™ Red In Situ Apoptosis Detection Kit
Shop All MilliporeSigma ProductsSpecifications
Specifications
| Description | ApopTag Red in situ apoptosis detection kit |
| For Use With (Application) | Immunohistochemistry (Paraffin), Immunocytochemistry, Immunohistochemistry |
| Includes | Equilibration Buffer, Reaction Buffer, TdT Enzyme, Stop/Wash Buffer, Blocking Solution, Anti-Digoxigenin-Rhodamine, Plastic Coverslips, affinity purified sheep polyclonal antibody |
| Quantity | 40 Assays |