Learn More
MilliporeSigma™ Chemicon™ beta-Integrin-mediated Cell Adhesion Array Kit, Colorimetric
Shop All MilliporeSigma ProductsSpecifications
Specifications
| Description | beta-Integrin-mediated Cell Adhesion Array Kit |
| Detection Method | Colorimetric |
| For Use With (Application) | For monitoring in vitro cell differentiation or genetic modification of cells |
| Includes | Assay Buffer, beta Integrin Array Plate, Cell Stain Solution, Extraction Buffer |
| Quantity | 1/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.