Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Chemicon™ BrdU Cell Proliferation Kits

Catalog No. 2750 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1000 Assays
200 Assays
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
27-50 200 Assays
27-52 1000 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 27-50 Supplier MilliporeSigma™ Supplier No. 2750

For ELISA

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Maintain the kit at 2-8°C under sterile conditions for up to 6 months after date of receipt
Description BrdU Cell Proliferation Kit
Detection Method Colorimetric
For Use With (Application) ELISA
Includes 50X Plate Wash Concentrate, BrdU Reagent, Fixing Solution, Prediluted BrdU Detection Antibody, Stop Solution, Goat anti-Mouse IgG, Peroxidase labeled, Conjugate Diluent, Substrate
Quantity 200 Assays
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.