Learn More
Specifications
Specifications
| Content And Storage | Maintain the kit at 2-8°C under sterile conditions for up to 6 months after date of receipt |
| Description | BrdU Cell Proliferation Kit |
| Detection Method | Colorimetric |
| For Use With (Application) | ELISA |
| Includes | 50X Plate Wash Concentrate, BrdU Reagent, Fixing Solution, Prediluted BrdU Detection Antibody, Stop Solution, Goat anti-Mouse IgG, Peroxidase labeled, Conjugate Diluent, Substrate |
| Quantity | 1000 Assays |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.