Learn More
MilliporeSigma™ Chemicon™ Caspase 3 Colorimetric Activity Assay Kit, DEVD
Shop All MilliporeSigma ProductsSpecifications
Specifications
| Content And Storage | -20°C |
| Description | Caspase 3 Colorimetric Activity Assay Kit, DEVD |
| Detection Method | Chromogenic |
| For Use With (Application) | Provide a simple and convenient means for assaying the activity of caspases that recognize the sequence DEVD |
| Includes | pNA Standard, 5X Cell Lysis Buffer, 5X Assay Buffer, Caspase-3 Substrate, Caspase-3 Inhibitor |
| Quantity | 25 Tests |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.