Learn More
MilliporeSigma™ Chemicon™ CaspaTag Pan-Caspase In Situ Assay Kit, Fluorescein
Shop All MilliporeSigma ProductsSpecifications
Specifications
| Content And Storage | 2°C to 8°C |
| Description | CaspaTag Pan-Caspase In Situ Assay Kit |
| Detection Method | Fluorescence |
| For Use With (Application) | For detection of active caspases in cells undergoing apoptosis |
| Includes | Hoechst Stain, FLICA Reagent, 10X Wash Buffer, Fixative, Propidium Iodide |
| Quantity | 25 Tests |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.