Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Chemicon™ Collagenase Type I

Click to view available options
Quantity:
250 mg
Description
Collagenase type I (from Clostridium histolyticum) is a crude collagenase preparation that can be used for the isolation of primary cells or for tissue dissociation by enzymatic means. The preparation also may contain caseinase, clostripain, and tryptic activities. This preparation can be used to dissociate cells in culture, and has been successfully used to passage human embryonic stem cells cultured with Millipore’s HEScGROTM Animal Component-Free Medium.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications
Specifications
Content And Storage | 2-8°C |
For Use With (Application) | Stem cell culture |
Quantity | 250 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction