Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Chemicon™ FibroGRO™ Xeno-Free Human Fibroblast Expansion Medium

Catalog No. p-5235453
Click to view available options
Quantity:
100 mL
500 mL

a new variation of our FibroGRO cell culture media that is optimized for the culture of fibroblasts without the use of animal-derived components.

FibroGRO™ Xeno-Free Human Fibroblast Expansion Medium is a new variation of our FibroGRO™ cell culture media that is optimized for the culture of fibroblasts without the use of animal-derived components. EMD Millipore’s FibroGRO™ cell system is perfect for establishing Xeno-Free human feeder layers for induced pluripotent, embryonic, and general stem cell culture. It can also be used to establish a model to study wound healing, toxicology and basic cell biology. FibroGRO™ Xeno Free supports the growth of these cells in a 2% human serum environment at rates equal to or greater than comparable media supplemented with 2-10% FBS. Our FibroGRO™ Xeno-Free media does not contain any antimicrobials, phenol red, or any other components that can cause cell stress and “masking effects” that may influence experimental results.
TRUSTED_SUSTAINABILITY

Specifications

Classification Xeno-free
For Use With (Application) Cell Culture
Quantity 100 mL
Content And Storage -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.