Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Chemicon™ Human Mesenchymal Stem Cell Kit
Shop All MilliporeSigma Products
Click to view available options
:
Human Mesenchymal Stem Cell Kit
Specifications
Specifications
Content And Storage | Store in liquid nitrogen |
For Use With (Application) | Stem cell culture |
Quantity | >1 million viable cells and 500mL of optimized media |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction