Learn More
MilliporeSigma™ Chemicon™ QCM Endothelial Cell Invasion Assay, 24-well, Colorimetric
Shop All MilliporeSigma ProductsSpecifications
Specifications
| Content And Storage | 2°C to 8°C |
| Description | Endothelial Cell Invasion Assay |
| Detection Method | Colorimetric |
| For Use With (Application) | Provides an in vitro model to quickly screen factors that can regulate endothelial invasion |
| Includes | Forceps, Cell Invasion Plate Assembly, Cell Stain Solution, Extraction Buffer, 24-well/96-Well plates, Swabs |
| Quantity | 1/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.