Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Chorionic Gonadotropin, Human Urine, Standard Grade, Protein

Catalog No. p-5364977
Click to view available options
Quantity:
1 mg
2.50 mg
Native, human chorionic gonadotropin from urine. Glycoprotein hormone synthesized by chorionic tissue of the placenta and found in urine during pregnancy. Also present in body fluids of patients with trophoblastic disease and ovarian tumors. Immunopotency: ≥3000IU/mg (WHO 1st IRP 75/551).
TRUSTED_SUSTAINABILITY

Specifications

Preparation Method H2O
Purification Method By weight
Quantity 1 mg
Immunogen Chorionic Gonadotropin
Storage Requirements Following reconstitution, aliquot and freeze (−20°C). Avoid freeze/thaw cycles of reconstituted solutions. Stock solutions are stable for up to 3 months at −20°C.
Regulatory Status RUO
Species Human
Recombinant Native
Form Solid
Purity or Quality Grade ≥30%, Standard Grade
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.