Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Complement C5a des-Arg, Human, Protein

Catalog No. 20490230UG Shop All MilliporeSigma Products
Click to view available options
Quantity:
30 μg
Native, human des-Arg C5a complement component. Single chain glycopeptide composed of 73 amino acids. Once the C5a glycopeptide has been produced in human plasma or serum, it is rapidly converted to C5a des-Arg upon removal of the C-terminal arginine by the endogenous serum carboxypeptidase N. Unlike the C3a des-Arg peptide, C5a des-Arg retains significant levels of its biological activities. Thus, C5a des-Arg has been shown to exhibit inflammatory cell chemotaxis, smooth muscle contraction, and leukotriene release from guinea pig lung.
TRUSTED_SUSTAINABILITY

Specifications

Concentration Concentration on Label
Formulation In 120mM NaCl, 10mM HEPES, pH 7.2.
Purification Method SDS-PAGE
Quantity 30 μg
Immunogen Complement C5a des-Arg
Storage Requirements Following initial thaw, aliquot and freeze (−70°C).
Regulatory Status RUO
Species Human
Recombinant Native
Form Liquid
Purity or Quality Grade ≥90%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.