Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ EmbryoMax™ ES Cell Qualified Fetal Bovine Serum
Click to view available options
Quantity:
100 mL
500 mL
Description
The EmbryoMax ES Cell Qualified, US origin Fetal Bovine Serum (FBS) is a growth supplement for cell culture media. Prescreened for stem cell culture
Specifications
Specifications
| Content And Storage | -20°C up to 2 years |
| Quantity | 100 mL |
| For Use With (Application) | Stem cell culture |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction