Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Endothelin 1, Human and Porcine, Polypeptide, Calbiochem
Click to view available options
Quantity:
0.1 mg
0.5 mg
1 mg
Description
A 21-amino acid polypeptide with potent vasoconstrictive action. Produces convulsions, arterial hypertension, and metabolic activation of periventricular white and gray matter. Its effects are mediated via L-type Ca2+ channels. Reported to activate phospholipase C in fibroblasts expressing ETA receptors.

Specifications
Specifications
Species | Human, Porcine |
Purification Method | HPLC |
Purity | ≥96% |
CAS | 117399-94-7 |
Color | White to Off-white |
Content And Storage | Following reconstitution, aliquot and freeze (−20°C). Stock solutions are stable for up to 2 weeks at −20°C. |
Format | Solid |
Formulation | Supplied as an acetate salt. |
Product Type | Endothelin 1 |
Quantity | 1 mg |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction