Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ ESGRO™ Chemicon™ 2i Supplement Kit (1000x)

Catalog No. p-5235533 Shop All MilliporeSigma Products
Click to view available options
Quantity:
1 Kit/5 L medium
1 Kit/500 mL medium

contains LIF a selective GSK3β and Mek 1/2 inhibitors to enhance viability of mouse embryonic stem and iPS cells in the absence of serum and feeder cells

The ESGRO™-2i Supplement Kit (1000x) contains LIF a selective GSK3β and Mek 1/2 inhibitors to enhance viability of mouse embryonic stem and iPS cells in the absence of serum and feeder cells. Supports a naive/ground pluripotent state.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage ESGRO Supplement: (Mfr. No. CS210511): Mouse recombinant Leukemia Inhibitory Factor (LIF), 0.5 x 106U/mL. One (1) 0.5mL vial. MEK/GS3 Inhibitor Supplement: (Mfr.. No. CS210510-500UL): One (1) 0.5mL vial. Store at -20°C up to 2 years
For Use With (Application) Stem cell culture
Quantity 1 Kit/500 mL medium
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.