Learn More
MilliporeSigma™ EXtrelut NT Prepacked Column: Filters
Catalog No. M1142360001 Shop All MilliporeSigma Products
Change view
Click to view available options
For Use With:
EXtrelut NT 20 HDPE Prepacked Columns
EXtrelut NT1 Prepacked Glass Columns
Packaging Quantity:
100 Units
50 Units
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With Packaging Quantity
M1142360001 EXtrelut NT1 Prepacked Glass Columns 100 Units
M1145670001 EXtrelut NT 20 HDPE Prepacked Columns 50 Units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1142360001 Supplier MilliporeSigma™ Supplier No. 1142360001
Add to Cart
Add to Cart
Spare filters for EXtrelut™ NT prepacked glass columns
Extrelut™ NT simplifies liquid-liquid extraction in its most effective form by using a chemically inert, wide-pore and highly pure diatomaceous earth based solid phase. Depending on the sample volume three types of columns are available: Extrelut™ NT1 can take up a maximum of one ml, Extrelut™ NT3 threemL and Extrelut™ NT20 up to twentymL of sample.
TRUSTED_SUSTAINABILITY
Specifications
Type Replacement Filter
Packaging Quantity 100 Units
For Use With EXtrelut NT1 Prepacked Glass Columns