Learn More
MilliporeSigma™ EZ-Fit™ Manifold for EZ-Fit™ Filtration Units
Catalog No. p-5063868
Click to view available options
Manifold Type:
1 place
For use in general microbiological analysis
  • Features specific EZ-Fit Filtration heads
  • Made from stainless steel
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
For Use With (Equipment) EZ-Fit™ Filtration Unit
Material Stainless Steel
Manifold Type 1 place
No. per Pack 1
Quantity 1/Pk.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.