Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Fibrinogen, Human Plasma, Protein
GSA_VA

Catalog No. p-5365063
Click to view available options
Quantity:
100 mg
1 g
Native fibrinogen from human plasma. A plasma glycoprotein, essential for clotting of blood, synthesized and secreted by hepatic parenchymal cells. Contains ≥90% clottable proteins.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from 20mM sodium citrate-HCl, pH 7.4.
Preparation Method Sterile dH2O
Purification Method Homogeneous by SDS-PAGE
Quantity 100 mg
Immunogen Fibrinogen
Storage Requirements Following reconstitution, aliquot and freeze (−70°C). Do not store solutions on ice; precipitation will occur. Stock solutions are stable for up to 6 months at −70°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Species Human
Recombinant Native
Form Lyophilized
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.