Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Glucagon, Human, Peptide hormone

Catalog No. 0523270001M
Click to view available options
Quantity:
0.1 mg
0.5 mg
Synthetic, human glucagon. Peptide hormone secreted by pancreatic α-cells. An important hyperglycemic agent.
TRUSTED_SUSTAINABILITY

Specifications

Species Human
Purification Method HPLC
Purity ≥97%
Content And Storage Following reconstitution, aliquot and freeze (−20°C). Stock solutions are stable up to 6 months at −20°C.
Format Lyophilized
Formulation Supplied as an acetate salt. Sold on the basis of peptide content.
Product Type Glucagon
Quantity 0.1 mg
Regulatory Status RUO - research use only
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.