Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Glucagon, Human, Peptide hormone
Click to view available options
Quantity:
0.5 mg
Description
Synthetic, human glucagon. Peptide hormone secreted by pancreatic α-cells. An important hyperglycemic agent.

Specifications
Specifications
Species | Human |
Purification Method | HPLC |
Purity | ≥97% |
Content And Storage | Following reconstitution, aliquot and freeze (−20°C). Stock solutions are stable up to 6 months at −20°C. |
Format | Lyophilized |
Formulation | Supplied as an acetate salt. Sold on the basis of peptide content. |
Product Type | Glucagon |
Quantity | 0.5 mg |
Regulatory Status | RUO - research use only |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction