Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Harleco™ APHA Wright Giemsa Solution

Catalog No. 23750078 Shop All MilliporeSigma Products
Click to view available options
Quantity:
1L
10L
This item is not returnable. View return policy
This item is not returnable. View return policy

For use with the MIDAS™ III Automated Stainer

  • Polychromic stain for peripheral blood and bone marrow smears
  • Used for red blood cell morphology and white blood cell differentiation
  • Specifically formulated for staining malarial parasites

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1L
Boiling Point 64.5° to 87.16°C
Format Liquid
Type Wright Giemsa Solution
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.