Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ HARLECO™ Gram Stain Reagents

Catalog No. p-103379 Shop All MilliporeSigma Products
Click to view available options
Quantity:
1 Set
4 x 500 mL
500 mL
Type:
Crystal Violet
Decolorizer
Gram Iodine
Gram Stain Kit
Safranin
This item is not returnable. View return policy

Catalog No. 23-750025

Supplier: MilliporeSigma™ 65092A95

This item is not returnable. View return policy

Complete set contains all reagents needed for Gram staining, or choose reagents individually.

 

  • Use qualitative technique for differential staining of bacteria from cultures and patient specimens by modified Hucker Gram stain method
  • Ideal for use with Midas III and other automated staining systems

 

Order Info

Individual reagents supplied in packs of four 500mL bottles

TRUSTED_SUSTAINABILITY

Specifications

Type Crystal Violet
Quantity 4 x 500 mL
Format Bottle
For Use With (Application) Gram staining of bacteria
Stain Type Crystal Violet
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.