Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Harleco™ Histology/Papanicolaou Stains: Gill Modified Hematoxylin

Catalog No. 23750017 Shop All MilliporeSigma Products
Change view
Click to view available options
Type:
Gill 1 Hematoxylin
Gill 2 Hematoxylin
Concentration:
1X
2X
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Concentration
23-750017 Gill 2 Hematoxylin 2X
23-750016 Gill 1 Hematoxylin 1X
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-750017 Supplier MilliporeSigma™ Supplier No. 6506685
This item is not returnable. View return policy

Harleco™ Histology/Papanicolaou Stains

TRUSTED_SUSTAINABILITY

Specifications

Concentration 2X
Type Gill 2 Hematoxylin
For Use With (Application) May be used for progressive or regressive staining in cytology and histology
Quantity 4000mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.