Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Harleco™ Histology/Papanicolaou Stains: Gill Modified Hematoxylin

Catalog No. p-4893245 Shop All MilliporeSigma Products
Click to view available options
Type:
Gill 1 Hematoxylin
Gill 2 Hematoxylin
Concentration:
1X
2X
This item is not returnable. View return policy
This item is not returnable. View return policy

Harleco™ Histology/Papanicolaou Stains

TRUSTED_SUSTAINABILITY

Specifications

Concentration 1X
Type Gill 1 Hematoxylin
For Use With (Application) May be used for progressive or regressive staining in cytology and histology
Quantity 4000mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.