Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Human Adipose Mesenchymal Stem Cells

Catalog No. scc038 Shop All MilliporeSigma Products
Click to view available options
Quantity:
1 x 106cells/vial

isolated from human adipose tissue and are cryopreserved at P2 to ensure optimal phenotype and the highest viability and plating efficiency.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in liquid nitrogen
For Use With (Application) Cell culture, cell differentiation, stem cell culture
Quantity 1 x 106cells/vial
Source Human adipose tissue
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.