Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Hyaluronic Acid Binding Protein, Bovine Nasal Cartilage, Biotinylated, Protein
Shop All MilliporeSigma Products
Click to view available options
:
50μg
Description
Affinity purified. Binds specifically and strongly to hyaluronic acid (≥2000 M.W.). Composed of two binding polypeptides, which are derived from N-terminal regions of the hyaluronic acid binding proteoglycan and linkage proteins. Useful for ELISA and histochemistry.
Specifications
Specifications
| Formulation | Lyophilized from 0.1M sodium bicarbonate solution, pH 8.3 |
| Preparation Method | Reconstitute with 100μL distilled H2O. |
| Quantity | 50μg |
| Immunogen | Hyaluronic Acid Binding Protein |
| Storage Requirements | Following reconstitution, aliquot and freeze (−20°C). Stock solutions are stable for up to 6 months at −20°C. |
| Regulatory Status | RUO |
| Conjugate | Biotin |
| Species | Bovine |
| Recombinant | Native |
| Form | Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction