Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Hyaluronic Acid Binding Protein, Bovine Nasal Cartilage, Protein

Catalog No. 385910100ug Shop All MilliporeSigma Products
Click to view available options
:
100μg

Catalog No. 38-591-0100UG

Supplier: MilliporeSigma™ 385910100UG

Only null left
Add to Cart
Add to Cart
Affinity purified. Binds specifically and strongly to hyaluronic acid (≥2000 M.W.). Composed of two binding polypeptides, which are derived from N-terminal regions of hyaluronic acid binding proteoglycan and linkage proteins.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from 0.1M Sodium Bicarbonate, pH 8.3.
Preparation Method Reconstitute in 100μL of PBS.
Quantity 100μg
Immunogen Hyaluronic Acid Binding Protein
Storage Requirements Following reconstitution, aliquot and freeze (−20°C). Stock solutions are stable for up to months at −20°C.
Regulatory Status RUO
Species Bovine
Recombinant Native
Form Lyophilized
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.