Specifications
| Description | ECM Cell Adhesion Assay |
| Detection Method | Fluorometric |
| Includes | Coated 96-Well Plate, Fluorescent Dye, D-PBS, and a user protocol |
| Quantity | 1/Pk. |
| Sample Type | Adherent Cultured Cells |